Assay Details
Amplicon Length: | 104 |
Chromosome Location: | Chr. 6 - 133740361 - 133740361 [-] on 133740361 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-370-5p | MIMAT0017202 |
Rat | rno-miR-370-3p | MIMAT0003122 |
Stem-loop Details
Rat
