Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | UUCUCGAGGAAAGAAGCACUUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-516a-5p | MIMAT0004770 |
| Gorilla gorilla | ggo-miR-516a-5p | MIMAT0036441 |
| Gorilla gorilla | ggo-miR-516a | MIMAT0036443 |
| Pongo pygmaeus | ppy-miR-516a-5p | MIMAT0015945 |
| Pongo pygmaeus | ppy-miR-516a | MIMAT0036453 |
Stem-loop Details
Human
Gorilla gorilla
Pongo pygmaeus

Genomic Map