Assay Details
Amplicon Length: | 145 |
Chromosome Location: | Chr. 19 - 53756391 - 53756391 [-] on 53756391 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-516a-5p | MIMAT0004770 |
| Human | hsa-miR-516a-3p | MIMAT0006778 |
Stem-loop Details
Human

Genomic Map