Assay Details
Gene Family ID: | MIPF0000204, mir-297 |
Mature miRNA Sequence: | UAUACAUACACACAUACCCAUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-297a-3p | MIMAT0004864 |
| Mouse | mmu-miR-297b-3p | MIMAT0004827 |
| Mouse | mmu-miR-297c-3p | MIMAT0004866 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-297a* (v17)
Mouse: mmu-miR-297c* (v17)
Mouse: mmu-miR-297c* (v17)

Genomic Map