Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 2 - 10516867 - 10516867 [-] on 10516867 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-297a-3p | MIMAT0004864 |
| Mouse | mmu-miR-297a-5p | MIMAT0000375 |
Stem-loop Details
Mouse

Genomic Map