Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | AAGUGCUUCCAUGUUUCAGUGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-302c-3p | MIMAT0003376 |
| Columba livia | cli-miR-302c-3p | MIMAT0038617 |
Stem-loop Details
Mouse
Columba livia

Genomic Map