Assay Details
Amplicon Length: | 93 |
Chromosome Location: | Chr. 3 - 127545095 - 127545095 [-] on 127545095 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-302a-5p | MIMAT0004579 |
| Mouse | mmu-miR-302a-3p | MIMAT0000380 |
| Mouse | mmu-miR-302b-5p | MIMAT0003373 |
| Mouse | mmu-miR-302b-3p | MIMAT0003374 |
| Mouse | mmu-miR-302c-5p | MIMAT0003375 |
| Mouse | mmu-miR-302c-3p | MIMAT0003376 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-mir-302a (v21)
Mouse: mmu-mir-302c (v21)
Mouse: mmu-mir-302c (v21)

Genomic Map