Assay Details
Gene Family ID: | MIPF0000070, mir-33 |
Mature miRNA Sequence: | CAGUGCCUCGGCAGUGCAGCCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-33b-3p | MIMAT0004811 |
| Pig | ssc-miR-33b-3p | MIMAT0044655 |
Stem-loop Details
Human
Pig

Genomic Map