Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 17 - 17814234 - 17814234 [+] on 17814234 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-33b-5p | MIMAT0003301 |
| Human | hsa-miR-33b-3p | MIMAT0004811 |
Stem-loop Details
Human

Genomic Map