Assay Details
Gene Family ID: | MIPF0001105, mir-3065 |
Mature miRNA Sequence: | UCAACAAAAUCACUGAUGCUGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3065-5p | MIMAT0015066 |
| Bovine | bta-miR-3065 | MIMAT0046741 |
Stem-loop Details
Human
Bovine

Genomic Map