Assay Details
Amplicon Length: | 48 |
Chromosome Location: | Chr. 17 - 81126383 - 81126383 [-] on 81126383 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3065-3p | MIMAT0015378 |
Human | hsa-miR-3065-5p | MIMAT0015066 |
Human | hsa-miR-338-3p | MIMAT0000763 |
Human | hsa-miR-338-5p | MIMAT0004701 |
Stem-loop Details
Human
