Assay Details
Gene Family ID: | MIPF0001105, mir-3065 |
Mature miRNA Sequence: | UCAGCACCAGGAUAUUGUUGGGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3065-3p | MIMAT0014837 |
| Horse | eca-miR-3065 | MIMAT0034658 |
Stem-loop Details
Mouse
Horse

Genomic Map