Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. 11 - 120015168 - 120015168 [-] on 120015168 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3065-5p | MIMAT0014836 |
| Mouse | mmu-miR-3065-3p | MIMAT0014837 |
| Mouse | mmu-miR-338-3p | MIMAT0000582 |
| Mouse | mmu-miR-338-5p | MIMAT0004647 |
Stem-loop Details
Mouse

Genomic Map