Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GUGGUUAUCCCUGCUGUGUUCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-487a-5p | MIMAT0026559 |
| Dasypus novemcinctus | dno-miR-487a-5p | MIMAT0047914 |
| Eptesicus fuscus | efu-miR-487 | MIMAT0035042 |
| Guinea pig | cpo-miR-487a-5p | MIMAT0047279 |
| Pteropus alecto | pal-miR-487a-5p | MIMAT0040375 |
Stem-loop Details
Human
Dasypus novemcinctus
Eptesicus fuscus
Guinea pig
Pteropus alecto

Genomic Map