Assay Details
Amplicon Length: | 65 |
Chromosome Location: | Chr. 14 - 101052247 - 101052247 [-] on 101052247 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-487a-5p | MIMAT0026559 |
| Human | hsa-miR-487a-3p | MIMAT0002178 |
Stem-loop Details
Human

Genomic Map