Assay Details
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | UGAGAUUCUCAUUCCUCCAUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ascaris suum | asu-miR-1175-3p | MIMAT0021534 |
Stem-loop Details
Ascaris suum
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | asu-miR-1175-3p |
|---|---|
| miRBase Accession Number: | MI0018586 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGAUUCUCAUUCCUCCAUUU |
| Species: | Ascaris suum |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | UGAGAUUCUCAUUCCUCCAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ascaris suum | asu-miR-1175-3p | MIMAT0021534 |