Assay Details
Gene Family ID: | MIPF0000715, mir-1175 |
Mature miRNA Sequence: | UGAGAUUCUCAUUCCUCCAUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ascaris suum | asu-miR-1175-3p | MIMAT0021534 |
Stem-loop Details
Ascaris suum
More Information
Related Products
miRBase Alias:
Atlantibacter subterranea: asu-miR-1175* (v18)

Genomic Map