Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | AUAAGACGAGCAAAAAGCUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-208a-3p | MIMAT0000241 |
| Mouse | mmu-miR-208a-3p | MIMAT0000520 |
| Anolis carolinensis | aca-miR-208-3p | MIMAT0021860 |
| Bovine | bta-miR-208a | MIMAT0009261 |
| Callithrix jacchus | cja-miR-208a | MIMAT0039597 |
| Dasypus novemcinctus | dno-miR-208a-3p | MIMAT0047751 |
| View More Rows | ||
| Dog | cfa-miR-208a | MIMAT0009868 |
| Guinea pig | cpo-miR-208a-3p | MIMAT0047106 |
| Horse | eca-miR-208a | MIMAT0012899 |
| Ophiophagus hannah | oha-miR-208-3p | MIMAT0036821 |
| Ornithorhynchus anatinus | oan-miR-208-3p | MIMAT0006921 |
| Pan troglodytes | ptr-miR-208a | MIMAT0008067 |
| Pongo pygmaeus | ppy-miR-208a | MIMAT0015799 |
| Pteropus alecto | pal-miR-208a-3p | MIMAT0040026 |
| Rabbit | ocu-miR-208a-3p | MIMAT0048330 |
| Rhesus monkey | mml-miR-208a-3p | MIMAT0006238 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-208a |
| Stem-loop Accession # | MI0000251 |
| Stem-loop Sequence | UGACGGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCA |
| Chromosome Location | Chr. 14 - 23388596 - 23388666 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000241
miRBase ID: hsa-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: Chr. 14 - 23388596 - 23388666 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-208a |
| Stem-loop Accession # | MI0000555 |
| Stem-loop Sequence | UUCCUUUGACGGGUGAGCUUUUGGCCCGGGUUAUACCUGACACUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCAGAGGAG |
| Chromosome Location | Chr. 14 - 54949060 - 54949142 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000520
miRBase ID: mmu-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: Chr. 14 - 54949060 - 54949142 [-] on Build GRCm38 |
| Stem-loop ID | aca-mir-208 |
| Stem-loop Accession # | MI0018811 |
| Stem-loop Sequence | GUUUCCUCUAACAAGGAAGCUUUUGGCUCGGGUUAUAUUUUAGAAUGGCAGUGUAUAAGACGAGCAAAAAGCUUGUUGGUUGGAGGAGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021860
miRBase ID: aca-miR-208-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | bta-mir-208a |
| Stem-loop Accession # | MI0009773 |
| Stem-loop Sequence | UGACGGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009261
miRBase ID: bta-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | cja-mir-208a-1 |
| Stem-loop Accession # | MI0032132 |
| Stem-loop Sequence | UUCCUGUGACGGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCAGAGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039597
miRBase ID: cja-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | cja-mir-208a-2 |
| Stem-loop Accession # | MI0032133 |
| Stem-loop Sequence | UCCUGUGACGGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCAGAGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039597
miRBase ID: cja-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | dno-mir-208a |
| Stem-loop Accession # | MI0039039 |
| Stem-loop Sequence | GAGCUUUUGGCCCGGGUUAUACCUGAUGUUCACGUAUAAGACGAGCAAAAAGCUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047751
miRBase ID: dno-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-208a |
| Stem-loop Accession # | MI0010365 |
| Stem-loop Sequence | ACGCAUGAGCUUUUGGCUCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009868
miRBase ID: cfa-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-208a |
| Stem-loop Accession # | MI0038700 |
| Stem-loop Sequence | GAGCUUUUGGCCCGGGUUAUACCUAAUGCUCACGUAUAAGACGAGCAAAAAGCUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047106
miRBase ID: cpo-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | eca-mir-208a |
| Stem-loop Accession # | MI0012655 |
| Stem-loop Sequence | UUCCUGUGACGGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCAGAGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012899
miRBase ID: eca-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | oha-mir-208 |
| Stem-loop Accession # | MI0031409 |
| Stem-loop Sequence | GGUGCUUCCUCUAAUGAAGAAGCUUUUGGCUCGGGUUAUAUUUGAACUCAACAGUGUAUAAGACGAGCAAAAAGCUUGUUGGUUGGAGGAGAUUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036821
miRBase ID: oha-miR-208-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | oan-mir-208 |
| Stem-loop Accession # | MI0006734 |
| Stem-loop Sequence | CGAGCUGGCCCUGGACCACUUCCUGUGACAGACAAGCUUUUGGCCCAGAUUAUACCUGACAUUCCCGUAUAAGACGAGCAAAAAGCUUGUUGGUCAGAGGAGCCACCAUCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006921
miRBase ID: oan-miR-208-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | ptr-mir-208a |
| Stem-loop Accession # | MI0008577 |
| Stem-loop Sequence | UGACGGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008067
miRBase ID: ptr-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-208a |
| Stem-loop Accession # | MI0014864 |
| Stem-loop Sequence | UGACAGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015799
miRBase ID: ppy-miR-208a Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | pal-mir-208a-1 |
| Stem-loop Accession # | MI0032454 |
| Stem-loop Sequence | GAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040026
miRBase ID: pal-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | pal-mir-208a-2 |
| Stem-loop Accession # | MI0032455 |
| Stem-loop Sequence | GAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040026
miRBase ID: pal-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-208a |
| Stem-loop Accession # | MI0039344 |
| Stem-loop Sequence | GAGCUUUUAGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048330
miRBase ID: ocu-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
| Stem-loop ID | mml-mir-208a |
| Stem-loop Accession # | MI0007668 |
| Stem-loop Sequence | UGACAGGCGAGCUUUUGGCCCGGGUUAUACCUGAUGCUCACGUAUAAGACGAGCAAAAAGCUUGUUGGUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006238
miRBase ID: mml-miR-208a-3p Mature miRNA Sequence: AUAAGACGAGCAAAAAGCUUGU Chromosome Location: NA |
More Information
Related Products

Genomic Map