Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 14 - 23388401 - 23388401 [-] on 23388401 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-208a-3p | MIMAT0000241 |
| Human | hsa-miR-208a-5p | MIMAT0026474 |
Stem-loop Details
Human

Genomic Map