Assay Details
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | UCAGUGCAUCACAGAACUUUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-148b-3p | MIMAT0000759 |
| Mouse | mmu-miR-148b-3p | MIMAT0000580 |
| Rat | rno-miR-148b-3p | MIMAT0000579 |
| Anolis carolinensis | aca-miR-148b-3p | MIMAT0021783 |
| Bovine | bta-miR-148b | MIMAT0003814 |
| Callithrix jacchus | cja-miR-148b | MIMAT0039462 |
| View More Rows | ||
| Chrysemys picta | cpi-miR-148b-3p | MIMAT0037784 |
| Cricetulus griseus | cgr-miR-148b-3p | MIMAT0023779 |
| Dasypus novemcinctus | dno-miR-148b-3p | MIMAT0047672 |
| Dog | cfa-miR-148b | MIMAT0006663 |
| Goat | chi-miR-148b-3p | MIMAT0035979 |
| Guinea pig | cpo-miR-148b-3p | MIMAT0047025 |
| Horse | eca-miR-148b-3p | MIMAT0012972 |
| Ophiophagus hannah | oha-miR-148b | MIMAT0036735 |
| Pan troglodytes | ptr-miR-148b | MIMAT0008042 |
| Pig | ssc-miR-148b-3p | MIMAT0013901 |
| Pongo pygmaeus | ppy-miR-148b | MIMAT0015772 |
| Pteropus alecto | pal-miR-148b-3p | MIMAT0039869 |
| Python bivittatus | pbv-miR-148b-3p | MIMAT0038967 |
| Rabbit | ocu-miR-148b-3p | MIMAT0048262 |
| Rhesus monkey | mml-miR-148b-3p | MIMAT0006209 |
| Tupaia chinensis | tch-miR-148b-3p | MIMAT0036534 |
| Xenopus tropicalis | xtr-miR-148b | MIMAT0003605 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-148b |
| Stem-loop Accession # | MI0000811 |
| Stem-loop Sequence | CAAGCACGAUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUA |
| Chromosome Location | Chr. 12 - 54337216 - 54337314 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000759
miRBase ID: hsa-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: Chr. 12 - 54337216 - 54337314 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-148b |
| Stem-loop Accession # | MI0000617 |
| Stem-loop Sequence | CAGGCACCCUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUA |
| Chromosome Location | Chr. 15 - 103285125 - 103285221 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000580
miRBase ID: mmu-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: Chr. 15 - 103285125 - 103285221 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-148b |
| Stem-loop Accession # | MI0000616 |
| Stem-loop Sequence | CAGGCACUCUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUA |
| Chromosome Location | Chr. 7 - 144912038 - 144912134 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000579
miRBase ID: rno-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: Chr. 7 - 144912038 - 144912134 [+] on Build Rnor_6.0 |
| Stem-loop ID | aca-mir-148b |
| Stem-loop Accession # | MI0018760 |
| Stem-loop Sequence | AGUUAAUGCAGCGCUUGAGGUGAAGUUCUGUUAUACACUCUGACUGUGGCUAUGUGGAAGUCAGUGCAUCACAGAACUUUGUCUCGAGAGCUUGCCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021783
miRBase ID: aca-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | bta-mir-148b |
| Stem-loop Accession # | MI0005030 |
| Stem-loop Sequence | UUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003814
miRBase ID: bta-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | cja-mir-148b |
| Stem-loop Accession # | MI0031986 |
| Stem-loop Sequence | AGCACUAUUAGCAUUUGAGGUGAAGUUCUGUUUUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUAGCAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039462
miRBase ID: cja-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | cpi-mir-148b |
| Stem-loop Accession # | MI0029494 |
| Stem-loop Sequence | AGCCCGCAGCAUUUGAGGUGAAGUUCUGUUAUACACUCUGACUGUGGCUACGUGGAGGUCAGUGCAUCACAGAACUUUGUCUCGAGAGCUUUUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037784
miRBase ID: cpi-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | cgr-mir-148b |
| Stem-loop Accession # | MI0020410 |
| Stem-loop Sequence | AUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023779
miRBase ID: cgr-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | dno-mir-148b |
| Stem-loop Accession # | MI0038998 |
| Stem-loop Sequence | GAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047672
miRBase ID: dno-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-148b |
| Stem-loop Accession # | MI0008069 |
| Stem-loop Sequence | GAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006663
miRBase ID: cfa-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | chi-mir-148b |
| Stem-loop Accession # | MI0030637 |
| Stem-loop Sequence | AUCAUUUCCAGUCACCGUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0035979
miRBase ID: chi-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-148b |
| Stem-loop Accession # | MI0038657 |
| Stem-loop Sequence | GAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047025
miRBase ID: cpo-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | eca-mir-148b |
| Stem-loop Accession # | MI0012724 |
| Stem-loop Sequence | AGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012972
miRBase ID: eca-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | oha-mir-148b |
| Stem-loop Accession # | MI0031362 |
| Stem-loop Sequence | GUUGUAGCGCUUGAGGUGAAGUUCUGUUAUACACUUUGACUGUGGCUACAUGGAAGUCAGUGCAUCACAGAACUUUGUCUCGAGAGCUUUUCAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036735
miRBase ID: oha-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | ptr-mir-148b |
| Stem-loop Accession # | MI0008548 |
| Stem-loop Sequence | CAAGCACGAUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008042
miRBase ID: ptr-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-148b |
| Stem-loop Accession # | MI0013118 |
| Stem-loop Sequence | AUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013901
miRBase ID: ssc-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-148b |
| Stem-loop Accession # | MI0014838 |
| Stem-loop Sequence | CAAGCACGAUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015772
miRBase ID: ppy-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | pal-mir-148b |
| Stem-loop Accession # | MI0032372 |
| Stem-loop Sequence | GAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039869
miRBase ID: pal-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-148b |
| Stem-loop Accession # | MI0030248 |
| Stem-loop Sequence | CUGUUACAGCGCUUGAGGUGAAGUUCUGUUAUACACUUUGACUGUGGCUACAUGGAAGUCAGUGCAUCACAGAACUUUGUCUCGAGAGCUUUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038967
miRBase ID: pbv-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-148b |
| Stem-loop Accession # | MI0039308 |
| Stem-loop Sequence | GAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048262
miRBase ID: ocu-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | mml-mir-148b |
| Stem-loop Accession # | MI0007639 |
| Stem-loop Sequence | CAAGCACGAUUAGCAUUUGAGGUGAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGUCUCGAAAGCUUUCUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006209
miRBase ID: mml-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | tch-mir-148b |
| Stem-loop Accession # | MI0031193 |
| Stem-loop Sequence | GAAGUUCUGUUAUACACUCAGGCUGUGGCUCUCUGAAAGUCAGUGCAUCACAGAACUUUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036534
miRBase ID: tch-miR-148b-3p Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
| Stem-loop ID | xtr-mir-148b |
| Stem-loop Accession # | MI0004845 |
| Stem-loop Sequence | UGAGGAACAGCGUUUGAGGUGAAGUUCUGUUAUACACUCCGGCUGUGAGUAAGUGGAAGUCAGUGCAUCACAGAACUUUGUCUCGAGGGCUUCACUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003605
miRBase ID: xtr-miR-148b Mature miRNA Sequence: UCAGUGCAUCACAGAACUUUGU Chromosome Location: NA |
More Information
Related Products

Genomic Map