Assay Details
Amplicon Length: | 53 |
Chromosome Location: | Chr. 15 - 103284829 - 103284829 [+] on 103284829 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-148b-3p | MIMAT0000580 |
Mouse | mmu-miR-148b-5p | MIMAT0017036 |
Stem-loop Details
Mouse
