Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | ACUUAAACGUGGAUGUACUUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302a-5p | MIMAT0000683 |
| Callithrix jacchus | cja-miR-302a-5p | MIMAT0039586 |
| Dasypus novemcinctus | dno-miR-302a-5p | MIMAT0047791 |
| Dog | cfa-miR-302a | MIMAT0009855 |
| Rabbit | ocu-miR-302a-5p | MIMAT0036320 |
Stem-loop Details
Human
Callithrix jacchus
Dasypus novemcinctus
Dog
Rabbit

Genomic Map