Assay Details
Gene Family ID: | MIPF0000468, mir-642 |
Mature miRNA Sequence: | GUCCCUCUCCAAAUGUGUCUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-642a-5p | MIMAT0003312 |
| Pan troglodytes | ptr-miR-642 | MIMAT0008296 |
| Rhesus monkey | mml-miR-642 | MIMAT0006483 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-642 (v15)
Human: hsa-miR-642a (v17)
Human: hsa-miR-642a (v17)

Genomic Map