Assay Details
Amplicon Length: | 91 |
Chromosome Location: | Chr. 19 - 45675230 - 45675230 [-] on 45675230 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-642a-5p | MIMAT0003312 |
| Human | hsa-miR-642a-3p | MIMAT0020924 |
| Human | hsa-miR-642b-5p | MIMAT0022736 |
| Human | hsa-miR-642b-3p | MIMAT0018444 |
Stem-loop Details
Human

Genomic Map