Assay Details
Gene Family ID: | MIPF0000193, mir-339 |
Mature miRNA Sequence: | UCCCUGUCCUCCAGGAGCUCACG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-339-5p | MIMAT0000764 |
| Mouse | mmu-miR-339-5p | MIMAT0000584 |
| Rat | rno-miR-339-5p | MIMAT0000583 |
| Cricetulus griseus | cgr-miR-339 | MIMAT0023919 |
| Dasypus novemcinctus | dno-miR-339-5p | MIMAT0047821 |
| Guinea pig | cpo-miR-339-5p | MIMAT0047176 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-339-5p | MIMAT0015842 |
| Rhesus monkey | mml-miR-339-5p | MIMAT0006279 |
Stem-loop Details
Human
Mouse
Rat
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Pongo pygmaeus
Rhesus monkey

Genomic Map