Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. 7 - 1022630 - 1022630 [-] on 1022630 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-339-3p | MIMAT0004702 |
| Human | hsa-miR-339-5p | MIMAT0000764 |
Stem-loop Details
Human

Genomic Map