Assay Details
Gene Family ID: | MIPF0000436, mir-544 |
Mature miRNA Sequence: | AUUCUGCAUUUUUAGCAAGCUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-544-3p | MIMAT0004941 |
Stem-loop Details
Mouse

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-544 (v15)
Rat: rno-miR-544 (v15)
Rat: rno-miR-544 (v15)
