Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 12 - 109729028 - 109729028 [+] on 109729028 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-544-3p | MIMAT0004941 |
Mouse | mmu-miR-544-5p | MIMAT0017282 |
Stem-loop Details
Mouse
