Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UAUUCAGGAAGGUGUUACUUAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-506-5p | MIMAT0022701 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-506-5p |
---|---|
miRBase Accession Number: | MI0003193 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAUUCAGGAAGGUGUUACUUAA |
Species: | Human |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UAUUCAGGAAGGUGUUACUUAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-506-5p | MIMAT0022701 |