Assay Details
Amplicon Length: | 98 |
Chromosome Location: | Chr. X - 147230417 - 147230417 [-] on 147230417 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-506-3p | MIMAT0002878 |
Human | hsa-miR-506-5p | MIMAT0022701 |
Stem-loop Details
Human
