Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. X - 134545923 - 134545923 [+] on 134545923 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-503-5p | MIMAT0002874 |
| Human | hsa-miR-503-3p | MIMAT0022925 |
Stem-loop Details
Human

Genomic Map