Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 1 - 117094942 - 117094942 [-] on 117094942 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-942-5p | MIMAT0004985 |
| Human | hsa-miR-942-3p | MIMAT0026734 |
Stem-loop Details
Human

Genomic Map