Assay Details
Gene Family ID: | MIPF0000511, mir-942 |
Mature miRNA Sequence: | UCUUCUCUGUUUUGGCCAUGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-942-5p | MIMAT0004985 |
| Pan troglodytes | ptr-miR-942 | MIMAT0008348 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map