Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. 12 - 111800057 - 111800057 [+] on 111800057 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6761-3p | MIMAT0027423 |
Human | hsa-miR-6761-5p | MIMAT0027422 |
Stem-loop Details
Human
