Assay Details
Mature miRNA Sequence: | UCCUACGCUGCUCUCUCACUCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6761-3p | MIMAT0027423 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6761-3p |
---|---|
miRBase Accession Number: | MI0022606 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCCUACGCUGCUCUCUCACUCC |
Species: | Human |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UCCUACGCUGCUCUCUCACUCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6761-3p | MIMAT0027423 |