Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UCGACAGCACGACACUGCCUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-196b-3p | MIMAT0009201 |
| Mouse | mmu-miR-196b-3p | MIMAT0017170 |
| Pteropus alecto | pal-miR-196b-3p | MIMAT0040010 |
Stem-loop Details
Human
Mouse
Pteropus alecto
More Information
Related Products
miRBase Alias:
Human: hsa-miR-196b* (v17)
Mouse: mmu-miR-196b* (v17)
Mouse: mmu-miR-196b* (v17)

Genomic Map