Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. 6 - 52230586 - 52230586 [+] on 52230586 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-196b-5p | MIMAT0001081 |
| Mouse | mmu-miR-196b-3p | MIMAT0017170 |
Stem-loop Details
Mouse

Genomic Map