Assay Details
Gene Family ID: | MIPF0000167, mir-370 |
Mature miRNA Sequence: | CAGGUCACGUCUCUGCAGUUACAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-370-5p | MIMAT0017202 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-370-5p |
---|---|
miRBase Accession Number: | MI0003486 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAGGUCACGUCUCUGCAGUUACAC |
Species: | Rat |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000167, mir-370 |
Mature miRNA Sequence: | CAGGUCACGUCUCUGCAGUUACAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-370-5p | MIMAT0017202 |