Assay Details
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | ACCUUGGCUCUAGACUGCUUACUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-212-5p | MIMAT0017158 |
| Ophiophagus hannah | oha-miR-212-5p | MIMAT0036828 |
Stem-loop Details
Rat
Ophiophagus hannah

Genomic Map