Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 10 - 62015249 - 62015249 [+] on 62015249 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-132-5p | MIMAT0017123 |
| Rat | rno-miR-132-3p | MIMAT0000838 |
| Rat | rno-miR-212-3p | MIMAT0000883 |
| Rat | rno-miR-212-5p | MIMAT0017158 |
Stem-loop Details
Rat

Genomic Map