Assay Details
Amplicon Length: | 100 |
Chromosome Location: | Chr. 16 - 77598741 - 77598741 [-] on 77598741 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-99a-5p | MIMAT0000131 |
| Mouse | mmu-miR-99a-3p | MIMAT0016981 |
Stem-loop Details
Mouse

Genomic Map