Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 12 - 109742112 - 109742112 [-] on 109742112 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-541-5p | MIMAT0003170 |
| Mouse | mmu-miR-541-3p | MIMAT0017209 |
Stem-loop Details
Mouse

Genomic Map