Assay Details
Gene Family ID: | MIPF0000213, mir-541 |
Mature miRNA Sequence: | AAGGGAUUCUGAUGUUGGUCACACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-541-5p | MIMAT0003170 |
| Rat | rno-miR-541-5p | MIMAT0003177 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-541 (v17)
Rat: rno-miR-541 (v18)
Rat: rno-miR-541 (v18)

Genomic Map