Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. X - 66789485 - 66789485 [+] on 66789485 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-883b-3p | MIMAT0004851 |
| Mouse | mmu-miR-883b-5p | MIMAT0004850 |
Stem-loop Details
Mouse

Genomic Map