Assay Details
Gene Family ID: | MIPF0000389, mir-883 |
Mature miRNA Sequence: | UAACUGCAACAUCUCUCAGUAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-883b-3p | MIMAT0004851 |
| Rat | rno-miR-883-3p | MIMAT0005290 |
Stem-loop Details
Mouse
Rat

Genomic Map