Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | UUUAACAUGGGGGUACCUGCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302c-5p | MIMAT0000716 |
| Dog | cfa-miR-302c | MIMAT0009857 |
| Rabbit | ocu-miR-302c-5p | MIMAT0036318 |
Stem-loop Details
Human
Dog
Rabbit

Genomic Map