Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCAUACAGGGACAUCCAGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-487a-3p | MIMAT0002178 |
| Callithrix jacchus | cja-miR-487a | MIMAT0039356 |
| Dasypus novemcinctus | dno-miR-487a-3p | MIMAT0047915 |
| Goat | chi-miR-487a-3p | MIMAT0036247 |
| Guinea pig | cpo-miR-487a-3p | MIMAT0047280 |
| Horse | eca-miR-487a | MIMAT0013161 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-487a | MIMAT0008161 |
| Pongo pygmaeus | ppy-miR-487a | MIMAT0015905 |
| Pteropus alecto | pal-miR-487a-3p | MIMAT0040376 |
| Rabbit | ocu-miR-487a-3p | MIMAT0048472 |
| Rhesus monkey | mml-miR-487a | MIMAT0006346 |
Stem-loop Details
Human
Callithrix jacchus
Dasypus novemcinctus
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-487 (v7.1)
Human: hsa-miR-487a (v19)
Human: hsa-miR-487a (v19)

Genomic Map