Assay Details
Gene Family ID: | MIPF0000070, mir-33 |
Mature miRNA Sequence: | GUGCAUUGCUGUUGCAUUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-33b-5p | MIMAT0003301 |
| Bovine | bta-miR-33b | MIMAT0009295 |
| Dog | cfa-miR-33b | MIMAT0009862 |
| Horse | eca-miR-33b | MIMAT0013037 |
| Pan troglodytes | ptr-miR-33b | MIMAT0008108 |
| Pig | ssc-miR-33b-5p | MIMAT0044654 |
| View More Rows | ||
| Pteropus alecto | pal-miR-33b-5p | MIMAT0040300 |
| Rhesus monkey | mml-miR-33b-5p | MIMAT0006173 |
Stem-loop Details
Human
Bovine
Dog
Horse
Pan troglodytes
Pig
Pteropus alecto
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-33b (v17)
Macaca mulatta: mml-miR-33b (v19)
Macaca mulatta: mml-miR-33b (v19)

Genomic Map