Assay Details
Gene Family ID: | MIPF0000038, mir-1 |
Mature miRNA Sequence: | UGGAAUGUAAAGAAGUGUGUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-1-3p | MIMAT0003125 |
| Cricetulus griseus | cgr-miR-1 | MIMAT0023725 |
| Guinea pig | cpo-miR-1b-3p | MIMAT0046836 |
Stem-loop Details
Rat
Cricetulus griseus
Guinea pig

Genomic Map