Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 18 - 2054783 - 2054783 [-] on 2054783 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-1-5p | MIMAT0003162 |
| Rat | rno-miR-1-3p | MIMAT0003125 |
Stem-loop Details
Rat

Genomic Map